Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014692.1 | NP_055507.1 | 11 | 1405 | ||
GenBank mRNA | AB007880.2 | - | 11 | 1405 |