Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037324.2 | NP_001032401.1 | 12, 13 | 1604 | ||
NM_001100120.1 | NP_001093590.1 | 13, 14 | 1829 | |||
NM_001100121.1 | NP_001093591.1 | 13, 14 | 1691 | |||
NM_014693.3 | NP_055508.3 | 13, 14 | 1940 | |||
GenBank mRNA | AB011176.1 | - | 13, 14 | 1686 | ||
AF192531.1 | - | 13, 14 | 1596 | |||
AF428263.1 | - | 13, 14 | 1868 | |||
AF428264.1 | - | 12, 13 | 1509 | |||
AF521189.1 | - | 13, 14 | 1805 | |||
AK293891.1 | - | 12, 13 | 1534 | |||
AK295270.1 | - | 6, 7 | 741 | |||
AK296541.1 | - | 12, 13 | 1587 | |||
AK309236.1 | - | 10, 11 | 1318 | |||
AY359003.1 | - | 12, 13 | 1560 | |||
BC142950.1 | - | 12, 13 | 1604 | 8860334 |