Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012431.2 | NP_036563.1 | 1 | 723 | ||
GenBank mRNA | AB002329.1 | - | 1 | 568 | ||
AK297209.1 | - | 1 | 403 | |||
BC140706.1 | - | 1 | 258 | 9021574 | ||
BC144338.1 | - | 1 | 258 | 9052858 | ||
BC152458.1 | - | 1 | 580 | 8862593 |