Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2bnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145726.1 | NP_001139198.1 | 6 | 600 | ||
GenBank mRNA | DV714338.1 | - | 6 | 583 |