Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099493.1 | NP_001092963.1 | 1 | 205 |
Set Membership: |
G-protein coupled receptor |