Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014706.3 | NP_055521.1 | 7, 8 | 1295 | ||
XM_005269241.4 | XP_005269298.1 | 7, 8 | 1362 | |||
XM_011539026.2 | XP_011537328.1 | 7, 8 | 830 | |||
GenBank mRNA | AB020880.1 | - | 7, 8 | 1079 | ||
AK225174.1 | - | 7, 8 | 1074 | |||
AK290209.1 | - | 7, 8 | 1087 | |||
AK299250.1 | - | 6, 7 | 786 | |||
AK302332.1 | - | 1, 2 | 50 | |||
AK304834.1 | - | 8, 9 | 997 | |||
BC032601.1 | - | 7, 8 | 1078 | |||
BC093784.1 | - | 7, 8 | 1065 | 7939629 | ||
BC103706.1 | - | 7, 8 | 1063 | |||
BC111983.1 | - | 7, 8 | 1065 | 8327451 | ||
BC143253.1 | - | 7, 8 | 1065 | 9051755 | ||
CR933631.1 | - | 1, 2 | 5357 | |||
D63879.1 | - | 7, 8 | 1060 |