Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014716.3 | NP_055531.1 | 6, 7 | 741 | ||
GenBank mRNA | BC018543.1 | - | 6, 7 | 716 | 4342840 | |
BT009788.1 | - | 6, 7 | 535 | |||
D30758.1 | - | 6, 7 | 723 |