Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ildr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769713.1 | XP_008767935.1 | 2 | 336 | ||
XM_008769714.1 | XP_008767936.1 | 2 | 275 | |||
XM_008769715.1 | XP_008767937.1 | 2 | 336 | |||
XM_008769716.1 | XP_008767938.1 | 2 | 2270 | |||
XM_008769717.1 | XP_008767939.1 | 2 | 2248 | |||
XM_008769718.1 | XP_008767940.1 | 2 | 140 | |||
XM_008769719.1 | XP_008767941.1 | 2 | 336 | |||
XM_008769720.1 | XP_008767942.1 | 2 | 336 |