Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC685482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763660.1 | XP_008761882.1 | 4 | 321 |
Gene Symbol: LOC298111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763616.1 | XP_008761838.1 | 4 | 321 |