Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014729.2 | NP_055544.1 | 1, 2 | 316 | ||
GenBank mRNA | AB018351.1 | - | 1, 2 | 318 | ||
AK000971.1 | - | 1, 2 | 98 | |||
AK302331.1 | - | 1, 2 | 240 | |||
BC016665.1 | - | 1, 2 | 159 | 4342384 |