Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228638.1 | XP_006228700.1 | 5 | 520 |
Gene Symbol: LOC100911249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759287.1 | XP_008757509.1 | 5 | 520 |