Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uroc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001066120.5 | XP_001066120.2 | 17 | 1716 | ||
XM_008763138.1 | XP_008761360.1 | 17 | 2143 |