Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143893.1 | NP_001137365.1 | 4 | 422 | ||
XM_006254426.2 | XP_006254488.1 | 4 | 2475 | |||
XM_008771955.1 | XP_008770177.1 | 4 | 2475 | |||
XM_008771956.1 | XP_008770178.1 | 3 | 275 | |||
GenBank mRNA | BC160837.1 | - | 4 | 406 |