Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ214601.1 | - | 8 | 950 |
Gene Symbol: LOC690183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001073579.4 | XP_001073579.1 | 1 | 942 |