Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014752.2 | NP_055567.2 | 4, 5 | 538 | ||
GenBank mRNA | AK293128.1 | - | 4, 5 | 519 | ||
BC008063.2 | - | 1 | 399 | 2967488 | ||
BC064957.1 | - | 4, 5 | 503 | 6092500 | ||
BC070276.1 | - | 1 | 276 | 4656138 | ||
BC082231.1 | - | 1 | 393 | 3954096 | ||
BC106066.1 | - | 4, 5 | 503 | 4521501 | ||
BP338843.1 | - | 4, 5 | 538 | |||
CR542233.1 | - | 2, 3 | 190 | |||
CR542243.1 | - | 2, 3 | 190 | |||
D14658.1 | - | 4, 5 | 497 |