Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pradc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749786.3 | XP_003749834.1 | 4 | 588 | ||
XM_008763097.1 | XP_008761319.1 | 4 | 604 |