Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031563.3 | NP_113751.3 | 8 | 1368 | ||
XM_008764058.1 | XP_008762280.1 | 8 | 1418 | |||
XM_008764059.1 | XP_008762281.1 | 7 | 1528 | |||
GenBank mRNA | BC072486.1 | - | 8 | 1366 | 7105037 | |
BC098672.1 | - | 8 | 1368 | 7124218 | ||
BC103637.1 | - | 8 | 1372 | 7107271 | ||
FQ232156.1 | - | 5 | 1008 | |||
M57299.1 | - | 8 | 1344 | |||
M69138.1 | - | 8 | 1503 |