Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqsec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236890.2 | XP_006236952.1 | 3 | 2178 | ||
XM_008763140.1 | XP_008761362.1 | 3 | 2178 | |||
XM_008763141.1 | XP_008761363.1 | 3 | 2178 |