Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136264.1 | NP_001129736.1 | 2 | 285 | ||
NM_001136266.1 | NP_001129738.1 | 2 | 285 | |||
NM_001136267.1 | NP_001129739.1 | 2 | 285 | |||
NM_001136268.1 | NP_001129740.1 | 2 | 285 | |||
NM_001136269.1 | NP_001129741.1 | 2 | 285 | |||
NM_014764.3 | NP_055579.1 | 2 | 285 | |||
GenBank mRNA | AK095963.1 | - | 2 | 144 | ||
AK125855.1 | - | 1 | 1523 | |||
AK290119.1 | - | 2 | 185 | |||
AK293330.1 | - | 2 | 183 | |||
AK303828.1 | - | 2 | 185 | |||
AY430097.1 | - | 2 | 184 | |||
BC002334.2 | - | 2 | 167 | 3509229 | ||
BC007900.2 | - | 2 | 166 | 4297784 | ||
BU662353.1 | - | 2 | 201 | |||
BX647278.1 | - | 2 | 183 | |||
CD697667.1 | - | 2 | 153 | |||
D31767.1 | - | 2 | 170 | |||
DA900264.1 | - | 2 | 180 | |||
DC303613.1 | - | 2 | 183 | |||
DC341917.1 | - | 2 | 286 |