Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134798.1 | NP_001128270.1 | 6 | 696 | ||
GenBank mRNA | FQ224263.1 | - | 6 | 709 |
Gene Symbol: NEWGENE_1582994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771413.1 | XP_008769635.1 | 6 | 864 |