Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SFI1 SIRNA Assays ›
Gene Symbol:
SFI1
Gene Name:
SFI1 centrin binding protein
Gene Aliases:
PISD, PPP1R139, hSfi1p
Chromosome Location:
Chr.22: 31496139 - 31618554 on Build GRCh38
UniGene ID:
Hs.62209
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18973
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SFI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007467.2
NP_001007468.1
3
717
NM_001258325.1
NP_001245254.1
3
717
NM_014775.3
NP_055590.2
3
717
XM_005261868.2
XP_005261925.1
3
715
XM_011530570.2
XP_011528872.1
3
715
XM_011530571.2
XP_011528873.1
3
711
XM_011530574.2
XP_011528876.1
3
392
XM_011530575.2
XP_011528877.1
3
715
XM_011530576.2
XP_011528878.1
3
715
XM_011530577.2
XP_011528879.1
3
711
XM_011530579.2
XP_011528881.1
3
392
XM_011530580.2
XP_011528882.1
3
715
XM_011530581.2
XP_011528883.1
3
392
XM_017029139.1
XP_016884628.1
3
392
XM_017029140.1
XP_016884629.1
3
715
XM_017029141.1
XP_016884630.1
3
392
XM_017029142.1
XP_016884631.1
3
715
XM_017029143.1
XP_016884632.1
3
711
XM_017029144.1
XP_016884633.1
3
715
XM_017029145.1
XP_016884634.1
3
715
XM_017029146.1
XP_016884635.1
3
392
XM_017029147.1
XP_016884636.1
3
715
XM_017029148.1
XP_016884637.1
3
711
XM_017029149.1
XP_016884638.1
3
715
XM_017029150.1
XP_016884639.1
3
711
XM_017029151.1
XP_016884640.1
3
711
XM_017029154.1
XP_016884643.1
3
711
XM_017029158.1
XP_016884647.1
3
711
XM_017029159.1
XP_016884648.1
3
392
XM_017029161.1
XP_016884650.1
2
218
XM_017029162.1
XP_016884651.1
2
218
GenBank mRNA
AB011114.2
-
3
581
AK292408.1
-
3
293
AK293987.1
-
3
330
BC129945.1
-
3
469
40124130
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
negative regulation of phosphatase activity
protein binding
phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0