Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173428.1 | NP_001166899.1 | 2 | 472 | ||
XM_006228478.2 | XP_006228540.1 | 2 | 467 | |||
XM_006228479.2 | XP_006228541.1 | 2 | 379 | |||
XM_006228480.2 | XP_006228542.1 | 2 | 467 | |||
XM_008758985.1 | XP_008757207.1 | 2 | 575 | |||
XM_008758986.1 | XP_008757208.1 | 2 | 577 | |||
XM_008758987.1 | XP_008757209.1 | 2 | 632 | |||
GenBank mRNA | BC168703.1 | - | 2 | 471 | 7455407 |