Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080496.2 | NP_001073965.2 | 2 | 158 | ||
GenBank mRNA | AK289566.1 | - | 2 | 151 | ||
BC001725.1 | - | 2 | 86 | 3533857 | ||
D87447.1 | - | 2 | 95 |