Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099487.1 | NP_001092957.1 | 5 | 1912 |
Gene Symbol: Vom2r38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099476.1 | NP_001092946.1 | 2131 |
Set Membership: |
G-protein coupled receptor |