Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234788.2 | XP_006234850.1 | 9 | 724 | ||
XM_006234789.2 | XP_006234851.1 | 9 | 704 | |||
XM_006234790.2 | XP_006234852.1 | 9 | 749 | |||
XM_006234791.2 | XP_006234853.1 | 8 | 606 | |||
XM_008762112.1 | XP_008760334.1 | 9 | 781 |