Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC24D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318066.1 | NP_001304995.1 | 15, 16 | 2306 | ||
NM_014822.3 | NP_055637.2 | 15, 16 | 2303 | |||
XM_005263379.2 | XP_005263436.1 | 15, 16 | 2180 | |||
XM_017008875.1 | XP_016864364.1 | 10, 11 | 1461 | |||
GenBank mRNA | AB018298.1 | - | 15, 16 | 2206 | ||
AF130464.2 | - | 14, 15 | 2006 | |||
AK001390.1 | - | 12, 13 | 1674 | |||
AK125962.1 | - | 9, 10 | 1294 | |||
AK291765.1 | - | 15, 16 | 2203 | |||
AK309960.1 | - | 8, 9 | 1042 | |||
BC035761.1 | - | 15, 16 | 2228 | 5554774 | ||
BC037736.1 | - | 18, 19 | 2780 |