Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H11A SIRNA Assays ›
Gene Symbol:
ZC3H11A
Gene Name:
zinc finger CCCH-type containing 11A
Gene Aliases:
ZC3HDC11A
Chromosome Location:
Chr.1: 203795537 - 203854128 on Build GRCh38
UniGene ID:
Hs.532399
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19127
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H11A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319238.1
NP_001306167.1
10
1817
NM_001319239.1
NP_001306168.1
10
1712
NM_014827.5
NP_055642.3
10
1388
XM_005245643.2
XP_005245700.1
10
1407
XM_005245644.3
XP_005245701.1
9
1091
XM_006711673.2
XP_006711736.1
10
1254
XM_011510199.1
XP_011508501.1
8
2459
XM_011510200.1
XP_011508502.1
9
1479
XM_011510201.1
XP_011508503.1
10
1665
XM_011510202.1
XP_011508504.1
8
1404
XM_017002956.1
XP_016858445.1
9
2713
XM_017002957.1
XP_016858446.1
9
2565
XM_017002958.1
XP_016858447.1
9
2552
XM_017002959.1
XP_016858448.1
10
1625
XM_017002960.1
XP_016858449.1
9
2713
XM_017002961.1
XP_016858450.1
8
2459
XM_017002962.1
XP_016858451.1
9
1479
XM_017002963.1
XP_016858452.1
10
1271
XM_017002964.1
XP_016858453.1
8
2459
XM_017002965.1
XP_016858454.1
7
1788
GenBank mRNA
AB014563.1
-
7
899
AK095858.1
-
10
1686
AK296983.1
-
6
746
BC014268.2
-
10
1226
3844154
BC038513.2
-
10
1543
BX648271.1
-
7
4201
BX649148.1
-
7
5003
CR627439.1
-
8
2409
CR627446.1
-
7
4512
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA processing factor
Gene Ontology Categories:
Function(s)
Process(es)
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
poly(A)+ mRNA export from nucleus
mRNA 3'-end processing
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0