Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135020.1 | NP_001128492.1 | 4 | 379 | ||
GenBank mRNA | BC168720.1 | - | 328 | 6922351 | ||
BC168779.1 | - | 4 | 349 | 6922409 | ||
CB313385.1 | - | 4 | 380 | |||
FQ221821.1 | - | 4 | 385 |