Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB007916.3 | - | 3 | 818 | ||
AK307886.1 | - | 2 | 482 | |||
CR936618.1 | - | 4 | 1018 |
Gene Symbol: SLC35E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199787.1 | NP_001186716.1 | 4 | 1218 | ||
NM_182838.2 | NP_878258.1 | 3 | 800 | |||
GenBank mRNA | AL049665.1 | - | 3 | 793 | ||
BC101666.1 | - | 2 | 399 | 8069172 | ||
BC112289.1 | - | 2 | 399 | 8327757 |