Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079712.1 | NP_001073180.1 | 399 | |||
GenBank mRNA | BC128760.1 | - | 2 | 399 | 7112650 |
Gene Symbol: Kdm4dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750432.1 | XP_003750480.1 | 2 | 798 |