Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134766.1 | NP_001128238.1 | 4 | 579 | ||
XM_006249355.2 | XP_006249417.1 | 4 | 1447 | |||
XM_006249357.2 | XP_006249419.1 | 4 | 1447 | |||
XM_006249358.1 | XP_006249420.1 | 4 | 520 | |||
XM_008769265.1 | XP_008767487.1 | 3 | 332 |