Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam91a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127578.1 | NP_001121050.1 | 24 | 3627 | ||
GenBank mRNA | BC166852.1 | - | 24 | 3627 | 9092514 |