Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014878.4 | NP_055693.4 | 4 | 472 | ||
GenBank mRNA | AK225953.1 | - | 4 | 437 | ||
AK292169.1 | - | 4 | 415 | |||
AK315531.1 | - | 4 | 451 | |||
AL832239.1 | - | 4 | 438 | |||
AL832245.1 | - | 4 | 435 | |||
BC016137.2 | - | 3 | 376 | 3908516 | ||
D13645.1 | - | 3 | 374 | |||
DA358022.1 | - | 4 | 472 |