Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014878.4 | NP_055693.4 | 6 | 662 | ||
GenBank mRNA | AF490254.1 | - | 3 | 146 | ||
AK225953.1 | - | 6 | 627 | |||
AK292169.1 | - | 6 | 605 | |||
AK315531.1 | - | 6 | 641 | |||
AL832239.1 | - | 6 | 628 | |||
AL832245.1 | - | 6 | 624 | |||
BC016137.2 | - | 5 | 566 | 3908516 | ||
CR456957.1 | - | 3 | 146 | |||
D13645.1 | - | 5 | 564 |