Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191101.1 | NP_001178030.1 | 36 | 4020 | ||
XM_006250582.2 | XP_006250644.1 | 38 | 4306 | |||
XM_008769943.1 | XP_008768165.1 | 38 | 4589 | |||
XM_008769944.1 | XP_008768166.1 | 39 | 4408 | |||
XM_008769945.1 | XP_008768167.1 | 37 | 5044 | |||
XM_008769946.1 | XP_008768168.1 | 36 | 4960 | |||
XM_008769947.1 | XP_008768169.1 | 36 | 4957 | |||
XM_008769948.1 | XP_008768170.1 | 36 | 4930 |