Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY75-CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198759.1 | NP_001185688.1 | 39 | 5538 | ||
NM_001198760.1 | NP_001185689.1 | 38 | 5370 | |||
GenBank mRNA | AY184222.1 | - | 39 | 5464 | ||
AY314006.1 | - | 38 | 5296 |
Gene Symbol: CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198763.1 | NP_001185692.1 | 5 | 424 | ||
NM_001198764.1 | NP_001185693.1 | 5 | 487 | |||
NM_014880.4 | NP_055695.2 | 6 | 598 | |||
GenBank mRNA | AF499000.1 | - | 6 | 552 | ||
AK291279.1 | - | 6 | 551 | |||
AK304422.1 | - | 5 | 411 | |||
AY314007.1 | - | 6 | 585 | |||
BC020646.1 | - | 5 | 434 | 4716449 | ||
D14664.1 | - | 6 | 539 | |||
DB460994.1 | - | 5 | 454 |