Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109569.1 | NP_001103039.1 | 8 | 801 | ||
XM_006230999.2 | XP_006231061.1 | 8 | 808 | |||
XM_006231000.2 | XP_006231062.1 | 8 | 676 | |||
GenBank mRNA | BC158834.1 | - | 8 | 770 | 7383724 |