Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191103.1 | NP_001178032.1 | 824 | |||
XM_008773612.1 | XP_008771834.1 | 929 |