Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005105.4 | NP_005096.1 | 4 | 420 | ||
GenBank mRNA | AF127761.1 | - | 4 | 322 | ||
AF161463.1 | - | 4 | 334 | |||
AF182415.1 | - | 4 | 339 | |||
AF198620.1 | - | 4 | 341 | |||
AF231511.1 | - | 4 | 327 | |||
AF299118.1 | - | 4 | 310 | |||
AK075009.1 | - | 4 | 341 | |||
BC017088.2 | - | 4 | 382 | 3850355 | ||
BC017770.1 | - | 2 | 660 | |||
BC071577.1 | - | 4 | 368 | |||
CR541805.1 | - | 4 | 310 | |||
CR541823.1 | - | 4 | 310 | |||
DB286556.1 | - | 4 | 420 | |||
KF051020.1 | - | 4 | 310 |