Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168641.1 | NP_001162112.1 | 6 | 1002 | ||
XM_006233028.2 | XP_006233090.1 | 5 | 2865 |