Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099490.1 | NP_001092960.1 | 185 |
Set Membership: |
G-protein coupled receptor |