Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105730.1 | NP_001099200.1 | 3 | 152 | ||
GenBank mRNA | FQ210333.1 | - | 3 | 144 | ||
FQ211223.1 | - | 3 | 144 | |||
FQ217403.1 | - | 3 | 149 | |||
FQ218290.1 | - | 3 | 143 | |||
FQ221642.1 | - | 3 | 144 | |||
FQ221735.1 | - | 3 | 145 | |||
FQ222008.1 | - | 3 | 144 | |||
FQ222172.1 | - | 3 | 144 | |||
FQ222320.1 | - | 3 | 144 | |||
FQ222499.1 | - | 3 | 144 | |||
FQ222594.1 | - | 3 | 147 | |||
FQ222598.1 | - | 3 | 145 | |||
FQ222664.1 | - | 3 | 144 | |||
FQ223680.1 | - | 3 | 145 | |||
FQ224357.1 | - | 3 | 144 | |||
FQ228386.1 | - | 3 | 145 | |||
FQ228826.1 | - | 3 | 143 | |||
FQ228989.1 | - | 3 | 144 | |||
FQ229601.1 | - | 3 | 144 | |||
X59277.1 | - | 3 | 133 |
Gene Symbol: LOC100359503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728428.3 | XP_002728474.1 | 1 | 149 |