Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759823.1 | XP_008758045.1 | 9 | 1382 | ||
XM_008759824.1 | XP_008758046.1 | 8 | 1087 |