Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100123.1 | NP_001093593.1 | 3 | 236 | ||
XM_006228169.2 | XP_006228231.1 | 2 | 283 | |||
XM_006228170.2 | XP_006228232.1 | 3 | 321 | |||
GenBank mRNA | AY998483.1 | - | 3 | 215 |