Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204344.1 | NP_001191273.1 | 1, 2 | 124 | ||
NM_005118.3 | NP_005109.2 | 3, 4 | 417 | |||
GenBank mRNA | AF520785.1 | - | 3, 4 | 423 | ||
AK291642.1 | - | 3, 4 | 328 | |||
AK313721.1 | - | 3, 4 | 392 | |||
AY434464.1 | - | 1, 2 | 124 | |||
BC069435.1 | - | 3, 4 | 372 | 7262219 | ||
BC074940.2 | - | 3, 4 | 341 | 30915494 | ||
BC074941.2 | - | 3, 4 | 341 | 30915271 | ||
BC104462.1 | - | 3, 4 | 416 | 40030800 | ||
BC104463.1 | - | 3, 4 | 417 | 40030803 |