Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099484.1 | NP_001092954.1 | 1965 |
Gene Symbol: Vom2r57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099474.1 | NP_001092944.1 | 6 | 1965 | ||
XM_008765199.1 | XP_008763421.1 | 7 | 1974 |
Set Membership: |
G-protein coupled receptor |