Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042494.1 | NP_001035959.1 | 4 | 680 | ||
NM_001042495.1 | NP_001035960.1 | 4 | 355 | |||
NM_001042496.1 | NP_001035961.1 | 4 | 746 | |||
NM_001042497.1 | NP_001035962.1 | 2 | 449 | |||
NM_005135.2 | NP_005126.1 | 3 | 319 | |||
NM_133647.1 | NP_598408.1 | 3 | 494 | |||
XM_006720793.3 | XP_006720856.1 | 3 | 1376 | |||
XM_011522269.2 | XP_011520571.1 | 3 | 1376 | |||
GenBank mRNA | AF105366.1 | - | 3 | 494 | ||
AF108831.1 | - | 3 | 228 | |||
AF116242.1 | - | 3 | 330 | |||
AF477977.1 | - | 4 | 687 | |||
AF531258.1 | - | 2 | 449 | |||
AF531259.1 | - | 4 | 687 | |||
AF531260.1 | - | 4 | 325 | |||
AK128133.1 | - | 3 | 149 | |||
AK292550.1 | - | 4 | 433 | |||
AK315283.1 | - | 3 | 291 | |||
BC051744.1 | - | 4 | 680 | 4829214 | ||
BC070107.1 | - | 4 | 350 | 4826538 | ||
BC126241.1 | - | 3 | 227 | 8991957 | ||
BC126243.1 | - | 3 | 227 | 8991959 | ||
BX648195.1 | - | 3 | 1436 | |||
DQ138323.1 | - | 4 | 745 |