Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173533.1 | NP_001167004.1 | 7 | 1272 | ||
NM_001173534.1 | NP_001167005.1 | 7 | 1263 | |||
NM_001184781.1 | NP_001171710.1 | 8 | 1386 | |||
NM_005137.2 | NP_005128.1 | 8 | 1395 | |||
NR_033674.1 | 7 | 1280 | ||||
GenBank mRNA | AK291670.1 | - | 8 | 1353 | ||
AK295697.1 | - | 7 | 1238 | |||
AK296337.1 | - | 7 | 1221 | |||
AK303130.1 | - | 8 | 1305 | |||
AK304382.1 | - | 7 | 1230 | |||
BC020339.1 | - | 8 | 1342 | |||
BC032430.1 | - | 8 | 1331 | 5243412 | ||
BC035425.1 | - | 8 | 1342 | |||
BC040500.1 | - | 8 | 1390 | 5272271 | ||
CR456433.1 | - | 8 | 1167 | |||
CR936871.1 | - | 9 | 1730 | |||
D79985.1 | - | 8 | 1332 | |||
L77560.1 | - | 8 | 1296 | |||
X83545.1 | - | 8 | 1296 | |||
X84076.1 | - | 8 | 1293 |