Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005258.2 | NP_005249.1 | 2, 3 | 224 | ||
GenBank mRNA | AK223430.1 | - | 2, 3 | 243 | ||
AK311872.1 | - | 2, 3 | 235 | |||
BC112262.1 | - | 2, 3 | 184 | 8327730 | ||
BC112264.1 | - | 2, 3 | 184 | 8327732 | ||
BC143904.1 | - | 2, 3 | 151 | 9052422 |