Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005331.4 | NP_005322.1 | 2 | 285 | ||
GenBank mRNA | BC056686.1 | - | 2 | 154 | 6738551 |